EMAIL THIS PAGE TO A FRIEND
The Journal of chemical physics
Mega three-photon absorption cross-section enhancement in pseudoisocyanine J-aggregates.
PMID 16392906
Abstract
Herein we report an extraordinary three-photon absorption cross-section (sigma'3) enhancement in J-aggregates supramolecular systems. The much higher value of sigma'3 in PIC J-aggregate (2.5 x 10(-71) cm6 s2 ph(-2)) compared to typical values obtained in organic molecules (10(-80) cm6 s2 ph(-2)) is attributed to the strong molecular transition dipole moment coupling in the supramolecular assembly. Three-photon absorption of PIC J-aggregates and monomer aqueous solutions were measured using the well known open aperture Z-scan technique pumping with a 25 ps pulse laser-OPG system at 1720 nm. This novel result opens new expectations for applications of supramolecular systems in bioimaging and medicine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
323764
1,1′-Diethyl-2,2′-cyanine iodide, 97%
C23H23IN2