EMAIL THIS PAGE TO A FRIEND
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes
Determination of acaricide residues in saudi arabian honey and beeswax using solid phase extraction and gas chromatography.
PMID 16393903
Abstract
Determination of acaricide residues of flumethrin, tau-fluvalinate, coumaphos, and amitraz in honey and beeswax was carried out using a rapid extraction method utilizing C-18 SPE cartridges and an analytical method utilizing GC with ECD, NPD, and MSD detectors for the four acaricides. Recovery percentages from the extraction method ranged from 90-102%, while the minimum detection levels ranged from 0.01-0.05 mg/kg for the acaricides. Nine of the 21 analyzed samples were found to be contaminated with the acaricides tau-fluvalinate and coumaphos. Neither flumethrin nor amitraz was detected in any of the honey or wax samples. Coumaphos was found only in honey samples in which two samples exceeded the tolerance levels set by EPA and EC regulations. It has not been detected in beeswax. Five honey samples and eight beeswax samples were found to be contaminated with tau-fluvalinate. One of the wax samples was contaminated with a relatively high residue of tau-fluvalinate and contained above 10 mg/kg.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46417
Flumethrin, PESTANAL®, analytical standard
C28H22Cl2FNO3