EMAIL THIS PAGE TO A FRIEND
Rapid communications in mass spectrometry : RCM
Enantiospecific separation and quantitation of mephenytoin and its metabolites nirvanol and 4'-hydroxymephenytoin in human plasma and urine by liquid chromatography/tandem mass spectrometry.
PMID 16395737
Abstract
A sensitive method using enantiospecific liquid chromatography/tandem mass spectrometry detection for the quantitation of S- and R-mephenytoin as well as its metabolites S- and R-nirvanol and S- and R-4'-hydroxymephenytoin in plasma and urine has been developed and validated. Plasma samples were prepared by protein precipitation with acetonitrile, while urine samples were diluted twice with the mobile phase before injection. The analytes were then separated on a chiral alpha(1)-acid glycoprotein (AGP) column and thereafter detected, using electrospray ionization tandem mass spectrometry. In plasma, the lower limit of quantification (LLOQ) was 1 ng/mL for S- and R-4'-hydroxymephenytoin and S-nirvanol and 3 ng/mL for R-nirvanol and S- and R-mephenytoin. In urine, the LLOQ was 3 ng/mL for all compounds. Resulting plasma and urine intra-day precision values (CV) were <12.4% and <6.4%, respectively, while plasma and urine accuracy values were 87.2-108.3% and 98.9-104.8% of the nominal values, respectively. The method was validated for plasma in the concentration ranges 1-500 ng/mL for S- and R-4'-hydroxymephenytoin, 1-1000 ng/mL for S-nirvanol, and 3-1500 ng/mL for R-nirvanol and S- and R-mephenytoin. The validated concentration range in urine was 3-5000 ng/mL for all compounds. By using this method, the metabolic activities of two human drug-metabolizing enzymes, cytochrome P450 (CYP) 2C19 and CYP2B6, were simultaneously characterized.
Related Materials