EMAIL THIS PAGE TO A FRIEND
Urological research
An unusual patient with kidney stones composed of 1-methyluric acid.
PMID 16397778
Abstract
An unusual case with kidney stones composed mainly of 1-methyluric acid is described. The patient, a Caucasian male of Celtic descent, reportedly drank at least eight cups of coffee per day and had a long history of rheumatoid arthritis, gouty attacks and renal colics--the latter attributed to nephrocalcinosis and analgesic nephropathy. He was treated with allopurinol. At 54 years, a bilateral nephrolithotomy was performed. Stone samples were analysed by thermogravimetry and infrared spectroscopy and reported to be 12-25% calcium oxalate, the remainder being organic uric acid-like material. Analysis of the extracts by HPLC confirmed that the organic material contained 67% of 1-methyluric acid and 33% of uric acid. Possible mechanisms leading to the precipitation of 1-methyluric acid from urine are discussed. We conclude that the high caffeine intake resulted in extremely elevated urinary concentrations of 1-methyluric acid favouring the formation of 1-methyluric acid stones.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6885
1-Methyluric acid
C6H6N4O3