EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Epoxide hydrolases: structure, function, mechanism, and assay.
PMID 16399371
Abstract
Epoxide hydrolases are a class of enzymes important in the detoxification of genotoxic compounds, as well as in the control of physiological signaling molecules. This chapter gives an overview on the function, structure, and enzymatic mechanism of structurally characterized epoxide hydrolases and describes selected assays for the quantification of epoxide hydrolase activity.