EMAIL THIS PAGE TO A FRIEND
American Journal of Clinical Nutrition
Ghrelin and glucagon-like peptide 1 concentrations, 24-h satiety, and energy and substrate metabolism during a high-protein diet and measured in a respiration chamber.
PMID 16400055