EMAIL THIS PAGE TO A FRIEND
Revue medicale de Liege
[The calcipotriol-betamethasone dipropionate association].
PMID 16402537
Abstract
The topical treatment of psoriasis benefits from the alternate use of dermocorticosteroids and vitamin D3 analogues. A new galenic formulation allows to combine them in a single application. Dovobet (LEO Pharma) ointment is the association of calcipotriol 50 microg/g with betamethasone dipoprionate 0.5 mg/g. This formulation boosts the therapeutic activity of calcipotriol. It also decreases the irritative inflammatory reaction due to calcipotriol without increasing the atrophogenic risk of the dermocorticoid.