EMAIL THIS PAGE TO A FRIEND
The Journal of nutrition
Medium-chain triacylglycerols enhance release of cholecystokinin in chicks.
PMID 1640264
Abstract
Whether medium-chain triacylglycerols (MCT) affect the plasma concentration of cholecystokinin (CCK) and crop-emptying rate in chicks was investigated after 0, 30, 60, 90, 120 or 180 min of diet intubation. Triacylglycerol sources used were corn oil [containing long-chain triacylglycerols (LCT)], glyceryl tricaprate and glyceryl tricaprylate at a level of 200 g/kg diet. Plasma CCK concentration was significantly enhanced in chicks given the two MCT treatments, but not in those given the LCT treatment, after 30 min feeding relative to the initial level. At all time points, chicks fed the diet containing LCT had significantly lower plasma CCK concentrations than those fed MCT, and chicks fed glyceryl tricaprate had higher concentrations than those fed glyceryl tricaprylate. Dietary MCT sources significantly delayed diet passage from the crop compared with dietary LCT. These results indicate that MCT are more potent stimulators of CCK secretion in chicks than LCT.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7517
Glyceryl tridecanoate, ≥99% (GC)
C33H62O6