EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Titanium(II)-mediated cyclizations of (silyloxy)enynes: a total synthesis of (-)-7-demethylpiericidin A1.
PMID 16402817
Abstract
A concise total synthesis of 7-demethylpiericidin A1 has been completed. The synthesis features a titanium(II)-mediated cyclization of a (silyloxy)enyne as the key step and proceeds in nine steps from tiglic aldehyde.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
192619
Tiglic aldehyde, ≥96%
C5H8O