EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Chemical synthesis and structure elucidation of bovine kappa-casein (1-44).
PMID 16403439
Abstract
The caseins (alphas1, alphas2, beta, and kappa) are phosphoproteins present in bovine milk that have been studied for over a century and whose structures remain obscure. Here we describe the chemical synthesis and structure elucidation of the N-terminal segment (1-44) of bovine kappa-casein, the protein which maintains the micellar structure of the caseins. kappa-Casein (1-44) was synthesised by highly optimised Boc solid-phase peptide chemistry and characterised by mass spectrometry. Structure elucidation was carried out by circular dichroism and nuclear magnetic resonance spectroscopy. CD analysis demonstrated that the segment was ill defined in aqueous medium but in 30% trifluoroethanol it exhibited considerable helical structure. Further, NMR analysis showed the presence of a helical segment containing 26 residues which extends from Pro8 to Arg34. This is the first report which demonstrates extensive secondary structure within the casein class of proteins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0406 κ-Casein from bovine milk, ≥70% (PAGE), lyophilized powder