EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Liquid chromatography-electrospray ionization mass spectrometry for direct identification and quantification of iophenoxic acid in serum.
PMID 16412709
Abstract
A liquid chromatographic-electrospray ionization mass spectrometric technique was developed for direct quantitation of iophenoxic acid (IA) in serum. IA was spiked into canine, feline, bovine, equine, and porcine sera, extracted, and quantified using negative ion monitoring following chromatographic separation on a Luna C18(2) 3 microm (100 mm x 2.1mm) reversed-phase column. The limit of detection was 25 ng/mL and the limit of quantification was 50 ng/mL. Inter- and intra-assay accuracy (86-113% and 87-115%, respectively) and precision (1.8-7.7%) were calculated. Analysis of serum collected from feral pigs, raccoons, and opossums following ingestion of IA-marked baits confirmed the appropriateness of this method for bait acceptance studies.