EMAIL THIS PAGE TO A FRIEND
Nihon rinsho. Japanese journal of clinical medicine
[Cytochrome P450 2C19 genotype -based second line therapy with amoxicillin and lansoprazole after failure of initial helicobacter pylori eradication therapy].
PMID 16416832