EMAIL THIS PAGE TO A FRIEND
Bioconjugate chemistry
Generic bioaffinity silicone surfaces.
PMID 16417248
Abstract
Synthetic polymer surfaces require surface modification to improve biocompatibility. A generic route to biocompatible silicone elastomers is described involving high yield surface functionalization of standard silicones with hydrosilanes, hydrosilylation using asymmetric, allyl-, NSC-terminated PEO of narrow molecular weight, and covalent modification in one step with amine-containing biological molecules including oligopeptides (YIGSR, RGDS), proteins (EGF, albumin, fibrinogen, mucin), and glycosaminoglycans (heparin). Efficient, high-density binding (e.g., 0.2 EGF molecules/nm2) was demonstrated using radiolabeling studies. The resulting surfaces were demonstrated to be biocompatible by further reaction with biomolecules, for example, thrombosis suppression on surfaces modified by heparin + ATIII, and the formation of confluent corneal epithelial cell layers on EGF, RGDS, or YIGSR surfaces.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7154
Tyr-Ile-Gly-Ser-Arg, ≥97% (HPLC)
C26H42N8O8