EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Measurement of [Ca2+] in cell suspensions using indo-1.
PMID 16422190
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
57180
Indo 1, for fluorescence, ≥80.0% (HPLC)
C32H26K5N3O12