EMAIL THIS PAGE TO A FRIEND
Cutis
Demodex mites as a cause of human disease.
PMID 16422462
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
257710
Selenium sulfide
S2Se