EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Simple method for the quantitative analysis of endogenous folate catabolites p-aminobenzoylglutamate (pABG) and its acetamido (apABG) derivative in human serum and urine by liquid chromatography-tandem mass spectrometry.
PMID 16427369
Abstract
To develop a routine method for quantitative measurement of the folate catabolites p-aminobenzoylglutamate (pABG) and acetamidobenzoylglutamate (apABG) in serum and urine using liquid chromatography-tandem mass spectrometry (LC-MS/MS). Urine, serum and aqueous standards were thawed. Two microliters of d3-glutamic acid (d3-Glu; 1 mmol/L) was added to 200 uL of specimen as internal standard. The samples were acidified with 4 uL 6N HCL, and aliquots were precipitated with 2 volumes (412 uL) of acetonitrile. For urine specimens 30 volumes (6.18 mL) of acetonitrile was used. Samples were centrifuged at 1900 x g for 10 min and the supernatant (10 microL) injected into a Biorad CAT/MET analytical column fitted to the LC-MS/MS. Detection of the catabolites was by selective multiple ion monitoring (multiple SRM) of the respective transitions. Urine and serum samples were analysed in a group of healthy volunteers and in anonymous samples from patients being tested for PTH and urinary catecholamines. pABG and apABG eluted at 5.2 and 4.74 min, respectively while the d3-glutamic acid eluted at around 7 min. Limit of quantitation (LOQ) for both catabolites was 10 nmol/L (which is equivalent to 33.3 fmol for a 10 microL injection). Limit of detection (LOD) was 1 nmol/L based on a signal to noise ratio of 5:1. A linear calibration curve was obtained from 10 to 100 nmol/L for serum specimens and from 10 to 200 micromol/L for urines. Imprecision for spiked serum samples (n=10) was between 2.5 and 20% for apABG and 4.5 and 21% for pABG (at 10 and 100 nmol/L, respectively). Imprecision for spiked urine samples (n=10) was between 2.9 and 4.0% for apABG and 6.0-12.7% for pABG. Recoveries were between 80 and 122% for serum samples and between 92 and 102% for urine specimens. Total folate catabolites in random urine samples from volunteers (n=5) are 2.9+/-2.3 umol/L (mean+/-S.D.). This group also had total serum catabolites of 11.9+/-7.6 nmol/L and serum folate of 35.3+/-5.8 nmol/L. Serum from patients being tested for PTH (n=11) had serum folate levels of 27.0+/-10.4 nmol/L with total serum catabolites of 20.4+/-23.8 nmol/L. Levels of serum folate and total catabolites in pregnant women (n=18) were 33.9+/-22.7 and 11.4+/-8.7 nmol/L, respectively. Mean urinary folate catabolites in patients being tested for urinary catecholamines (n=19) was 581.8+/-368.4 nmol/L. A simple, reliable and highly specific method by LC-MS/MS for detecting and quantifying the folate catabolites pABG and apABG was developed. This enables, for the first time, the routine clinical analysis of folate utilization in patients.