EMAIL THIS PAGE TO A FRIEND
Chemico-biological interactions
BW284c51 blocks nicotinic acetylcholine receptors transplanted to Xenopus oocytes.
PMID 16429545
Abstract
We have studied the effects of BW284c51 on the function of Torpedo nicotinic acetylcholine (Ach) receptors (nAchRs) transplanted to Xenopus laevis oocytes. BW284c51 reversible inhibited Ach-elicited currents (IAch) in a concentration-dependent manner, increased IAch desensitisation and changed the Ach concentration-dependence of the IAch from a two-site to a single-site Hill equation, without affecting the EC50. These effects were only present at hyperpolarising potentials, suggesting that nAchR blockade by BW284c51 is non-competitive and likely due to an open channel block as the principal mechanism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9013
1,5-Bis(4-allyldimethylammoniumphenyl)pentan-3-one dibromide
C27H38Br2N2O