EMAIL THIS PAGE TO A FRIEND
Drug metabolism and disposition: the biological fate of chemicals
Interactions of amoxicillin and cefaclor with human renal organic anion and peptide transporters.
PMID 16434549
Abstract
Amoxicillin and cefaclor are two of the widely used beta-lactam antibiotics in the treatment of urinary tract infections. Both drugs are eliminated mainly by the kidney and rely on renal excretion to exert their antibacterial activities in the urinary tract. Previous studies have suggested the involvement of organic anion and oligopeptide transporters in membrane transport of beta-lactams. The objective of the current study was to examine the kinetics of amoxicillin and cefaclor interactions with human renal transporters human organic anion transporter 1 (hOAT1), human peptide transporter 1 (hPepT1), and human peptide transporter 2 (hPepT2) in detail, both as substrates and as inhibitors. Using fluorescence protein tagging and cell sorting, we established Madin-Darby canine kidney cell lines stably expressing highly functional hOAT1, hPepT1, and hPepT2. Amoxicillin and cefaclor inhibited hOAT1-mediated [(3)H]para-aminohippuric acid uptake (K(i) = 11.0 and 1.15 mM, respectively). However, our uptake study revealed that neither drug was transported by hOAT1. Amoxicillin and cefaclor competitively inhibited hPepT2-mediated [(3)H]glycylsarcosine uptake (K(i) = 733 and 65 muM, respectively), whereas much lower affinity for hPepT1 was observed with both antibiotics. Direct uptake studies demonstrated that amoxicillin and cefaclor were transported by hPepT1 and hPepT2. Kinetic analysis showed that hPepT2-mediated uptake of both drugs was saturable with K(m) of 1.04 mM for amoxicillin and 70.2 muM for cefaclor. hPepT2, and to a lesser extent hPepT1, may play an important role in apical transport of amoxicillin and cefaclor in the renal tubule. hOAT1, in contrast, is not involved in basolateral uptake of these antibiotics.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6895
Cefaclor
C15H14ClN3O4S