EMAIL THIS PAGE TO A FRIEND
Journal of neuroscience research
Acetoacetate protects neuronal cells from oxidative glutamate toxicity.
PMID 16435389
Abstract
Glutamate cytotoxicity contributes to neuronal degeneration in many central nervous system (CNS) diseases, such as epilepsy and ischemia. We previously reported that a high-fat and low-carbohydrate diet, the ketogenic diet (KD), protects against kainic acid-induced hippocampal cell death in mice. We hypothesized based on these findings that ketosis resulting from KD might inhibit glutamate cytotoxicity, resulting in inhibition of hippocampal neuronal cell death. Therefore, we investigated the role of ketone bodies [acetoacetate (AA) and beta-hydroxybutyrate (beta-OHB)] both in a mouse hippocampal cell line (HT22) and in rat primary hippocampal neurons. As a result, we found that pretreatment with 5 mM lithium AA and 4 mM Na beta-OHB protected the HT22 hippocampal cell line and primary hippocampal neuronal culture against 5 mM glutamate toxicity and that up to 2 hr of pretreatment with 5 mM AA had a protective effect against 5 mM glutamate toxicity in the HT22 cell line. Pretreatment with 5 mM AA decreased ROS production of HT22 cell line at 2 and 8 hr exposure of glutamate, and it decreased the appearance of annexin V-positive HT22 cells, which are indicative of an early stage of apoptosis, and propidium iodide-positive HT22 cells, which are indicative of necrosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
54925
(S)-3-Hydroxybutyric acid, ≥97.0%
C4H8O3