EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Mechanistic insights into triterpene synthesis from quantum mechanical calculations. Detection of systematic errors in B3LYP cyclization energies.
PMID 16446812
Abstract
Most quantum mechanical studies of triterpene synthesis have been done on small models. We calculated mPW1PW91/6-311+G(2d,p)//B3LYP/6-31G* energies for many C30H51O+ intermediates to establish the first comprehensive energy profiles for the cationic cyclization of oxidosqualene to lanosterol, lupeol, and hopen-3beta-ol. Differences among these 3 profiles were attributed to ring strain, steric effects, and proton affinity. Modest activation energy barriers and the ample exothermicity of most annulations indicated that the cationic intermediates rarely need enzymatic stabilization. The course of reaction is guided by hyperconjugation of the carbocationic 2p orbital with parallel C-C and C-H bonds. Hyperconjugation for cations with a horizontal 2p orbital (in the plane of the ABCD ring system) leads to annulation and ring expansion. If the 2p orbital becomes vertical, hyperconjugation fosters 1,2-methyl and hydride shifts. Transition states leading to rings D and E were bridged cyclopropane/carbonium ions, which allow ring expansion/annulation to bypass formation of undesirable anti-Markovnikov cations. Similar bridged species are also involved in many cation rearrangements. Our calculations revealed systematic errors in DFT cyclization energies. A spectacular example was the B3LYP/6-311+G(2d,p)//B3LYP/6-31G* prediction of endothermicity for the strongly exothermic cyclization of squalene to hopene. DFT cyclization energies for the 6-311+G(2d,p) basis set ranged from reasonable accuracy (mPW1PW91, TPSSh with 25% HF exchange) to underestimation (B3LYP, HCTH, TPSS, O3LYP) or overestimation (MP2, MPW1K, PBE1PBE). Despite minor inaccuracies, B3LYP/6-31G* geometries usually gave credible mPW1PW91 single-point energies. Nevertheless, DFT energies should be used cautiously until broadly reliable methods are established.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
04626
Hop-22(29)-ene solution, 0.1 mg/mL in isooctane, analytical standard
C30H50