EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Resolving the motional modes that code for RNA adaptation.
PMID 16456078
Abstract
Using a domain elongation strategy, we decoupled internal motions in RNA from overall rotational diffusion. This allowed us to site-specifically resolve a manifold of motional modes in two regulatory RNAs from HIV-1 with the use of nuclear magnetic resonance spin relaxation methods. Base and sugar librations vary on a picosecond time scale and occur within helical domains that move collectively at diffusion-limited nanosecond time scales. Pivot points are short, functionally important, and highly mobile internal loops. These spontaneous changes in RNA conformation correlate quantitatively with those that follow adaptive recognition of diverse targets. Thus, ligands may stabilize existing RNA conformations rather than inducing new ones.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3913
L-Argininamide dihydrochloride
C6H15N5O · 2HCl