EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Spectroscopic and thermal studies of chromium(III), molybdenum(VI) and ruthenium(0) complexes of maleic hydrazide.
PMID 16458051
Abstract
Interaction of maleic hydrazide (LH(2)) with [Cr(CO)(6)] in air at atmospheric pressure resulted in the formation of the complex [(LH)Cr(mu-O)(2)Cr(LH)] (1). Reaction of LH(2) with [Mo(CO)(6)] in air also gave the complex [(LH(2))O(2)Mo(mu-O)(2)MoO(2)(LH(2))] (2). Under the same conditions, the reaction of LH(2) with [Ru(3)(CO)(12)] resulted in the formation of the tricarbonyl complex [Ru(CO)(3)(LH(2))] (3). The complexes were characterized by elemental analysis, IR, and (1)H NMR spectroscopy. The thermal properties of the complexes were investigated by thermogravimetry technique.
Related Materials