EMAIL THIS PAGE TO A FRIEND
Journal of immunological methods
Measurement of high affinity antibodies on antigen-immunoblots.
PMID 16458918
Abstract
We describe a semi-quantitative method for measuring the relative affinity of antigen-specific oligoclonal IgG bands separated by isoelectric focusing followed by blotting onto antigen-coated membrane and incubation with sodium thiocyanate. When the developed blot is digitised in greyscale, densitograms can be made and peak areas calculated using ImageJ freeware. By expressing peak area as a percentage of the total area under the curve we have shown that there is a statistically significant rise in percentage of peak area for a given band which persists with increasing molarities of sodium thiocyanate.