EMAIL THIS PAGE TO A FRIEND
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan
[Reaction of pyridinium salts with active methylene compounds].
PMID 16462100
Abstract
The carbon-carbon bond-forming reactions of 1-methyl-2-methylthiophenylpyridinium iodides with active methylene compounds (such as dimethyl malonate, malononitrile) using NaH as a base gave the 2-substituted methylene-1,2-dihydropyridine derivatives.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
136441
Dimethyl malonate, 98%
C5H8O4
63380
Dimethyl malonate, purum, ≥96.0% (GC)
C5H8O4