EMAIL THIS PAGE TO A FRIEND
The Yale journal of biology and medicine
Myocardial infarction after taking zolmitriptan.
PMID 16464313
Abstract
We report the case of a patient with mild non-obstructive coronary artery disease who sustained an inferior wall myocardial infarction shortly after taking zolmitriptan as abortive therapy for migraine headaches. A Medline search was performed to review all reported cases of myocardial infarction related to migraine therapy with zolmitriptan and related medications. Zolmitriptan may cause myocardial infarction (MI) even in the absence of significant coronary artery disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0248
Zolmitriptan, ≥98% (HPLC)
C16H21N3O2