EMAIL THIS PAGE TO A FRIEND
Gender medicine
The negative impact of litigation on women's health care.
PMID 16464731
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7909
Dicyclomine hydrochloride, ≥99% (TLC), powder
C19H35NO2 · HCl