EMAIL THIS PAGE TO A FRIEND
European journal of cardiovascular nursing : journal of the Working Group on Cardiovascular Nursing of the European Society of Cardiology
Metabolic therapeutics in angina pectoris: history revisited with perhexiline.
PMID 16469541
Abstract
The ever-increasing burden of ischaemic heart disease and its common manifestation chronic angina pectoris calls for the exploration of other treatment options for those patients who despite the maximum conventional pharmacological and surgical interventions continue to suffer. Such exploration has led to the increasing use of new metabolically acting antianginal agents and the re-emergence of an old and somewhat forgotten pharmacological agent, perhexiline maleate. This review aims to update the cardiac nurse with knowledge to manage the care a patient receiving perhexiline maleate treatment and provide a brief review of three new metabolic agents: trimetazidine, ranolazine and etomoxir.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0120
Perhexiline maleate salt, ≥98% (HPLC)
C19H35N · C4H4O4