EMAIL THIS PAGE TO A FRIEND
Chemphyschem : a European journal of chemical physics and physical chemistry
Conformational landscapes and free-jet rotational spectrum of indan-1-ol.
PMID 16470643
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
328413
(R)-(−)-1-Indanol, 99%
C9H10O
323128
(S)-(+)-1-Indanol, 99%
C9H10O
193739
1-Indanol, 98%
C9H10O