EMAIL THIS PAGE TO A FRIEND
Toxicology in vitro : an international journal published in association with BIBRA
Differential induction of apoptosis by type A and B trichothecenes in Jurkat T-lymphocytes.
PMID 16472964
Abstract
Several studies have shown that the mycotoxins T-2 toxin, diacetoxyscirpenol (DAS), deoxynivalenol (DON) and nivalenol (NIV) affect lymphocyte functioning. However, the molecular mechanisms underlying the immunomodulatory effects of these trichothecenes are not defined yet. In this study, the potency of the type A trichothecenes T-2 toxin and DAS, and the type B trichothecenes DON (and its metabolite de-epoxy-deoxynivalenol; DOM-1) and NIV to reduce mitochondrial activity and to induce apoptosis of Jurkat T cells (human T lymphocytes) were examined. T-2 toxin and DAS are much more cytotoxic at low concentrations than DON and NIV as shown by the AlamarBlue cytotoxicity assay. In addition, the mechanism whereby DON and NIV induced cytotoxicity is mainly via apoptosis as we observed phosphatidylserine externalization, mitochondrial release of cytochrome c, procaspase-3 degradation and Bcl-2 degradation. In contrast, type A trichothecenes reduce the mitochondrial activity at approximately 1000-fold lower concentrations than the type B trichothecenes, resulting in necrosis. These data suggest that the mechanisms resulting in cytotoxic effects are different for type A and type B trichothecenes.