EMAIL THIS PAGE TO A FRIEND
Journal of Pediatric Surgery
An association of multiple endocrine neoplasia 2B, a RET mutation; constipation; and low substance P-nerve fiber density in colonic circular muscle.
PMID 16481266