EMAIL THIS PAGE TO A FRIEND
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes
Biotransformation of metamitron by human p450 expressed in transgenic tobacco cell cultures.
PMID 16484082
Abstract
In the present investigation, the oxidative metabolism of 14C-labeled metamitron was examined in plant cell cultures of tobacco overexpressing human P450 enzymes CYP1A1 or CYP1A2; special interest was in the aromatic hydroxylation of the herbicide. The oxidative metabolites deaminometamitron (DAM) and 4-hydroxydeaminometamitron (4-HDAM) were found in the untransformed control culture as well as in the transgenic culture. The transgenic cultures, however, exhibited higher turnover rates after 48 h of incubation with 20 microg 14C-metamitron per assay (untransformed: 40%, CYP1A1: 80%, CYP1A2: 100%). Primary metabolite 4-HDAM was partially found in glucosylated form in the transgenic cultures. As minor oxidative metabolites, 6-hydroxyphenyl-3-methoxymethyl-1,2,4-triazine-5(4H)-one and 3-hydroxymethyl-6-phenyl-1,2,4-triazine-5(4H)-one were identified in the transgenic cultures by GC-MS, LC-MS. Additionally, it could be demonstrated that both foreign enzymes (CYP1A1, CYP1A2) also catalyzed the deamination of metamitron. In a large-scale study (up to 400 microg per assay) with the transgenic culture expressing CYP1A2, the high efficiency of this P450 system toward metamitron was demonstrated: turnover of the xenobiotic was almost complete with 400 microg. Since large portions of unglucosylated 4-H-DAM were found, the activity of foreign CYP1A2 apparently exceeded that of endogenous O-glucosyltransferases of the tobacco cell culture. We concluded that in comparison to the nontransformed cell culture, the extent of metabolism was considerably higher in the transgenic cultures. The transgenic cell cultures expressing human CYP1A1 or CYP1A2 are thus suitable tools for the production of large quantities of primary oxidized metabolites of metamitron.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36154
Metamitron, PESTANAL®, analytical standard
C10H10N4O