EMAIL THIS PAGE TO A FRIEND
Critical care medicine
Effects of therapeutic bronchoalveolar lavage and partial liquid ventilation on meconium-aspirated newborn piglets.
PMID 16484898
Abstract
To investigate the therapeutic effects of bronchoalveolar lavage (BAL) with either diluted surfactant (SBAL) or perfluorochemical liquid (PBAL), followed by either conventional mechanical ventilation (CMV) or partial liquid ventilation (PLV), on lung injury and proinflammatory cytokine production induced by meconium aspiration in newborn piglets. A prospective, randomized, experimental study. An animal research laboratory at a medical center. Anesthetized and mechanically ventilated newborn piglets (n = 27). The animals were instilled with 3-5 mL/kg 25% human meconium via an endotracheal tube to induce meconium aspiration syndrome (MAS). After stabilization, animals were randomly assigned to either CMV group (no BAL) or one of the treatment groups (SBAL-CMV, SBAL-PLV, PBAL-CMV, and PBAL-PLV). Cardiopulmonary variables were monitored, and interleukin-1beta and interleukin-6 content of the serum and lung tissue was measured. The animals without any treatment (CMV group) displayed the worst outcome; the animals in the PBAL-PLV group had the best gas exchange, lung compliance, and least pulmonary damage; and the SBAL-CMV, PBAL-CMV, and SBAL-PLV groups had intermediate effects. The serum interleukin-1beta concentration of the CMV group was significantly higher than all other groups over time (p < .05), and interleukin-6 concentration was significantly higher than the PBAL-PLV group (p < .05). The tissue interleukin-1beta and interleukin-6 contents were also highest in the CMV group and lowest in the PBAL-PLV group. Initial therapeutic BAL and therapeutic BAL followed by PLV with the same perfluorochemical liquid provided significant therapeutic effects in treating an animal model with severe MAS and therefore warrant consideration in cases that are intractable to other therapies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
302937
Perfluoro(methylcyclohexane), technical grade, 90%
C7F14