EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
Arylamine-induced hemolytic anemia: electron spin resonance spectrometry studies.
PMID 1648864
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
671576
N-Phenylhydroxylamine, ≥97.0% (GC)
C6H7NO