EMAIL THIS PAGE TO A FRIEND
Applied microbiology and biotechnology
Coupling of enantioselective biooxidation of DL-1,2-propanediol and bioreduction of pinacolone via regeneration cycle of coenzyme.
PMID 16489454
Abstract
Enantioselective biotransformation of DL-1,2-propanediol to D-2-hydroxypropanic acid was first reported by the authors. In the biooxidation process, there were some by-product formed and thus influenced the e.e. value and output of the acid. Restricting oxygen in the reaction system and offering additional proton receptor to the system displayed approving effect. The latter method constructed regeneration cycle system of coenzyme. In the article, the bioreduction of pinacolone was coupled to the enantioselective oxidation. Yield of the acid was increased by 36% and e.e. value of the product approached 99%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P45605
3,3-Dimethyl-2-butanone, 98%
C6H12O