EMAIL THIS PAGE TO A FRIEND
Biotechnology letters
Biosynthesis of medium-chain-length poly(hydroxyalkanoates) from soy molasses.
PMID 16489492
Abstract
Pseudomonas corrugata was selected from a screening process for the bioconversion of inexpensive soy molasses into medium-chain-length poly(hydroxyalkanoates) (mcl-PHA). We obtained yields of 1.5 g cell dry weight (CDW)/l culture with growth medium supplemented with 2% (w/v) soy molasses, and of an average of 3.4 g CDW/l with 5% (w/v) soy molasses. Crude PHAs were obtained at 5-17% of CDW. The most prominent repeat-unit monomers in the PHAs were 3-hydroxydodecanoate, 3-hydroxyoctanoate, 3-hydroxydodecanoate, and 3-hydroxytetradecenoate. This work represents the first description of fermentative mcl-PHA production from the soy molasses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H3398
DL-β-Hydroxylauric acid, ≥99% (GC)
C12H24O3