EMAIL THIS PAGE TO A FRIEND
Pflugers Archiv : European journal of physiology
Increased resistance to fatigue in creatine kinase deficient muscle is not due to improved contractile economy.
PMID 16491397
Abstract
There has been speculation on the origin of the increased endurance of skeletal muscles in creatine kinase (CK)-deficient mice. Important factors that have been raised include the documented increased mitochondrial capacity and alterations in myosin heavy chain (MyHC) isoform composition in CK-deficient muscle. More recently, the absence of inorganic phosphate release from phosphocreatine hydrolysis in exercising CK-deficient muscle has been postulated to contribute to the lower fatigueability in skeletal muscle. In this study, we tested the hypothesis that the reported shift in MyHC composition to slower isoforms in CK-deficient muscle leads to a decrease in oxygen cost of twitch performance. To that aim, extensor digitorum longus (EDL) and soleus (SOL) muscles were isolated from wild-type (WT) and knock-out mice deficient in the cytoplasmic muscle-type and sarcomeric mitochondrial isoenzymes of CK, and oxygen consumption per twitch time-tension-integral (TTI) was measured. The results show that the adaptive response to loss of CK function does not involve any major change to contractile economy of skeletal muscle.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G6878
3-Guanidinopropionic acid
C4H9N3O2