EMAIL THIS PAGE TO A FRIEND
Seikagaku. The Journal of Japanese Biochemical Society
[Discovery and recent development of a novel coenzyme pyrroloquinoline quinone (PQQ)].
PMID 1649236
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7783
Pyrroloquinoline quinone, ≥95.0% (HPLC)
C14H6N2O8
64682
Pyrroloquinoline quinone, ≥97.0% (HPLC)
C14H6N2O8