EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Oxidative stress promotes polarization of human T cell differentiation toward a T helper 2 phenotype.
PMID 16493032
Abstract
These studies were conducted to determine the effects of oxidative stress on human T cell differentiation and polarization into Th1 or Th2 phenotypes. Highly purified naive CD4+ T cells were isolated from PBMC of healthy, nonatopic donors. CD4+ T cells were stimulated with anti-CD3 and anti-CD28 mAb in the presence or absence of oxidative stress as supplied by 2,3-dimethoxy-1,4-naphthoquinone (DMNQ), which generates a low level of superoxide anion. Increases in cellular superoxide were observed by exposure to DMNQ. Exposure of unpolarized CD4+ T cells to IL-12 or IL-4 resulted in a Th1 or Th2 phenotype, respectively. T cells stimulated in the absence of polarizing cytokines secreted modest amounts of IFN-gamma and TNF-alpha. Cells stimulated in the continuous presence of 5 microM DMNQ, displayed a marked up-regulation in Th2 cytokines, including IL-4, IL-5, and IL-13, but not the Th1 cytokine IFN-gamma. Th2 responses were blunted by concomitant exposure to thiol antioxidants. Long-term exposure of T cells to DMNQ resulted in growth of cells expressing CCR4, and a decrease in cells expressing CXCR3, indicating phenotypic conversion to Th2 cells. These results suggest that oxidative stress favors a Th2-polarizing condition.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5439 2,3-Dimethoxy-1,4-naphthoquinone, ≥99%, solid
C12H10O4