EMAIL THIS PAGE TO A FRIEND
Proteomics
A mass spectrometry compatible silver staining method for protein incorporating a new silver sensitizer in sodium dodecyl sulfate-polyacrylamide electrophoresis gels.
PMID 16493709
Abstract
A quick, sensitive, and MALDI-TOF MS compatible silver staining method, namely Eriochrome black T (EBT)-silver method, is described. The method can detect 0.05-0.2 ng protein within 60 min in SDS-PAGE gels. EBT dye was used as a silver ion sensitizer having reducing power for silver ions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
858390
Eriochrome® Black T, ACS reagent (indicator grade)
C20H12N3NaO7S