EMAIL THIS PAGE TO A FRIEND
Environmental toxicology and chemistry
Biochemical and spectroscopic studies of the response of Convolvulus arvensis L. to chromium(III) and chromium(VI) stress.
PMID 16494245
Abstract
The objective of the present study was to determine the oxidative stress caused by hexavalent chromium (Cr[VI]), the chromium (Cr) uptake, and the Cr speciation in Convolvulus arvensis L. plants grown in hydroponics media containing either Cr(VI) or Cr(III). The results demonstrated that C. arvensis plants exposed to Cr(VI) concentrations ranging from 0 to 40 mg/L expressed higher ascorbate peroxidase specific activity in roots than in shoots. On the other hand, catalase activity monitored in plants exposed to 2 mg/L of Cr(VI) for 24 h increased in roots after a few hours of exposure. However, catalase activity in shoots revealed a decrement almost immediately after treatment was initiated. The results from x-ray absorption spectroscopic studies indicated that the oxidation state of the supplied Cr(III) remained the same in plant tissues. The supplied Cr(VI), however, was reduced to the trivalent form in plant tissues. The results of inductively coupled plasma/optical emission spectroscopy demonstrated that after 5 d, the roots of plants exposed to 40 mg/L of Cr(III) or Cr(VI) accumulated approximately 25,000 and 3,500 mg/kg dry weight of Cr, respectively. Nevertheless, shoots concentrated 1,500 and 2,000 mg/kg dry weight of Cr from Cr(III) and Cr(VI), respectively, which indicated that Cr moved faster into C. arvensis plants when supplied as Cr(VI).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
239259
Chromium(III) nitrate nonahydrate, 99%
CrN3O9 · 9H2O