EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Novel synthesis of desymmetrized resorcinol derivatives: aryl fluoride displacement on deactivated substrates.
PMID 16497012
Abstract
A short, high-yielding synthesis of differentially substituted resorcinol derivatives has been developed that utilizes 1,3-difluorobenzene as the starting material and employs sequential nucleophilic aromatic substitution (S(N)Ar) reactions to generate desymmetrized products. The scope and limitations of the second S(N)Ar reaction on the deactivated 1-alkoxy-3-fluorobenzene intermediates have been investigated. This methodology has also been employed in the synthesis of desymmetrized catechol derivatives from 1,2-difluorobenzene.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
126152
1,2-Difluorobenzene, 98%
C6H4F2
D102008
1,3-Difluorobenzene, ≥99%
C6H4F2