EMAIL THIS PAGE TO A FRIEND
FEBS letters
Fumarate reductase: structural and mechanistic insights from the catalytic reduction of 2-methylfumarate.
PMID 16497301
Abstract
The soluble fumarate reductase (FR) from Shewanella frigidimarina can catalyse the reduction of 2-methylfumarate with a k(cat) of 9.0 s(-1) and a K(M) of 32 microM. This produces the chiral molecule 2-methylsuccinate. Here, we present the structure of FR to a resolution of 1.5 A with 2-methylfumarate bound at the active site. The mode of binding of 2-methylfumarate allows us to predict the stereochemistry of the product as (S)-2-methylsuccinate. To test this prediction we have analysed the product stereochemistry by circular dichroism spectroscopy and confirmed the production of (S)-2-methylsuccinate.
Related Materials