EMAIL THIS PAGE TO A FRIEND
Expert opinion on emerging drugs
Emerging drug therapies for benign prostatic hyperplasia.
PMID 16503830
Abstract
Benign prostatic hyperplasia (BPH) is the nonmalignant enlargement of the prostate gland caused by increases in number of both epithelial and stromal cells. Clinically, BPH leads to voiding dysfunction, which is most often referred to as lower urinary tract symptoms (LUTS). Historically, the only treatments for LUTS due to BPH were watchful waiting or surgery (transurethral or open prostatectomy). However, over the last 20 years medical therapy has taken a prominent role in the management of BPH. Current medical treatments for BPH include alpha-adrenergic receptor antagonists, inhibitors of the 5-alpha reductase enzyme and various phytotherapies. These agents are generally effective and safe; however, many patients are unable to tolerate the side effects or are refractory to medical management and require surgery. In light of this, many potential new therapies for the treatment of BPH are under development. Some represent a variation of current treatments, whereas others target novel molecular pathways within the prostate. The aim of this review is to examine current pharmacotherapies as well as to highlight emerging drugs that may improve our treatment of patients with LUTS secondary to BPH.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L4900
Lonidamine, mitochondrial hexokinase inhibitor
C15H10Cl2N2O2