EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Pre-treatment of high oil and grease pet food industrial wastewaters using immobilized lipase hydrolyzation.
PMID 16504384
Abstract
Wastewaters generating from pet food industries contain high concentration of oil and grease (O&G), which is difficult to treat through conventional biological treatment systems. In this study, the hydrolysis of O&G originating from pet food industrial wastewater was evaluated. Candida rugosa lipase was immobilized in calcium alginate beads and applied in the hydrolysis experiment. Results showed that approximately 50% of the O&G was hydrolyzed due to the enzyme activity. A significant increment in COD and VFA production was also observed. The immobilized lipase activity was confirmed with p-nitrophenyl palmitate (pNPP) before and after O&G hydrolysis. During the 3-day experiment, approximately 65% of the beads were recovered and after the hydrolysis, approximately 70% of the enzyme activity remained in the beads. This study shows the potential of immobilized lipase as a pre-treatment step in biological treatment of pet food manufacturing wastewater.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N2752
4-Nitrophenyl palmitate, lipase substrate
C22H35NO4