EMAIL THIS PAGE TO A FRIEND
International journal of toxicology
Involvement of calcium in 7beta -hydroxycholesterol and cholesterol-5beta,6beta -epoxide-induced apoptosis.
PMID 16510355
Abstract
Oxidized low-density lipoprotein (oxLDL) is believed to play a central role in the development of atherosclerosis. The induction of apoptosis in cells of the arterial wall is a critical event in the development of atheroma. 7beta-Hydroxycholesterol (7beta-OH) and cholesterol-5beta,6beta-epoxide (beta-epoxide) are components of oxLDL and have previously been shown to be potent inducers of apoptosis. The exact mechanism through which these oxysterols induce apoptosis remains to be fully elucidated. A perturbation of intracellular calcium homeostasis has been found to trigger apoptosis in many experimental systems. The aim of the present study was to determine the involvement of calcium signaling in 7beta-OH and beta-epoxide-induced apoptosis. To this end, the authors employed the calcium channel blockers verapamil and nifedipine and inhibitors of calpain activation, ALLM and ALLN. Verapamil protected against the decrease in viability induced by 7beta-OH whereas nifedipine had a protective effect in both 7beta-OH and beta-epoxide-treated cells, though these compounds did not restore viability to control levels. Verapamil, nifedipine, and ALLM prevented apoptosis induced by beta-epoxide. None of the compounds employed in the current study protected against 7beta-OH-induced apoptosis. Our results implicate calcium signaling in the apoptotic pathway induced by beta-epoxide and also highlight differences between apoptosis induced by 7beta-OH and beta-epoxide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2773
Cholesterol 5α,6α-epoxide
C27H46O2
C2648
Cholesterol 5β,6β-epoxide, ≥98%
C27H46O2