EMAIL THIS PAGE TO A FRIEND
Acta crystallographica. Section F, Structural biology and crystallization communications
Expression, purification and crystallization of Streptococcus dysgalactiae-derived mitogen.
PMID 16511312
Abstract
Superantigens are bacterial or viral toxins with potent immunostimulatory properties. Streptococcus dysgalactiae-derived mitogen, a 25 kDa protein, is a recently discovered superantigen isolated from S. dysgalactiae culture supernatant. Sequence considerations suggest that it belongs to a new superantigen family distinct from other superantigens. The protein was expressed in Escherichia coli cells and purified to homogeneity. Crystals were grown at pH 4.2-4.4 in the presence of 18-20%(w/v) PEG 3350 and 0.4 M lithium nitrate. A complete data set to 2.4 A resolution was collected from a single crystal at liquid-nitrogen temperatures using synchrotron radiation. The crystals belong to space group P3/P3(1)/P3(2), with unit-cell parameters a = b = 52.7, c = 62.4 A, gamma = 120 degrees and one molecule in the crystallographic asymmetric unit.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
62574
Lithium nitrate, BioUltra, ≥99.0% (calc. on dried substances, T)
LiNO3