EMAIL THIS PAGE TO A FRIEND
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry
A highly sensitive and rapid assay for protein determination in human urine and penicillin using a Rayleigh light-scattering technique with benzeneazo-8-acetylamino-1-naphthol-3,6-disulfonic acid sodium salt.
PMID 16512422
Abstract
A simple and sensitive method was conducted for the determination of trace amounts of proteins with benzeneazo-8-acetylamino-1-naphthol-3,6-disulfonic acid sodium salt (azophloxine, AP) using a Rayleigh light-scattering (RLS) technique. At pH 2.60 and in the presence of an emulsifier OP microemulsion, the RLS of AP can be greatly enhanced by proteins, owing to the interaction between AP and protein. The enhanced intensity is proportional to the concentration of proteins. Four proteins, including bovine serum albumin (BSA), human serum albumin (HSA), lysozyme (Lys) and gamma globulin (gamma-G) have been tested. For example, the linear range of BSA was 0 - 0.06 microg mL(-1) with detection limits of 2.38 ng mL(-1). The method was applied to the analysis of protein in human urine and penicillin samples with satisfactory results. The relative standard deviation was in all instances less than 4.0%, and the recovery was in the range of 97.5 - 104%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
210633
Acid Red 1, Dye content 60 %
C18H13N3Na2O8S2
11640
Azophloxine, for microscopy (Hist.)
C18H13N3Na2O8S2
40462
Red 2G, analytical standard
C18H13N3Na2O8S2