EMAIL THIS PAGE TO A FRIEND
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry
Spectrophotometric determination of anthracycline anticancer agents with aluminum(III) and chromazurol S in a nonionic surfactant micellar medium.
PMID 16512429
Abstract
A simple and highly sensitive spectrophotometric method for the determination of anthracycline anticancer agents, such as Daunorubicin hydrochloride (DAU), was established by using aluminum(III) and Chromazurol S (CAS) in a nonionic surfactant micellar medium. In the case of determination of DAU, the apparent molar absorptivity was 1.3 x 10(5) dm3 mol(-1) cm(-1) at 615 nm. Beer's law was obeyed in the concentration range of 0.028 - 2.82 microg ml(-1) for DAU. Owing to no need for solvent extraction, this method could be applied to assays of DAU and related drugs in pharmaceutical preparations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
199532
Chromeazurol S, Dye content 50 %
C23H13Cl2Na3O9S