EMAIL THIS PAGE TO A FRIEND
Water research
Biodegradation of oxadiazon by a soil isolated Pseudomonas fluorescens strain CG5: Implementation in an herbicide removal reactor and modelling.
PMID 16516265
Abstract
An oxadiazon-degrading bacterial, Pseudomonas strain CG5, was isolated from an agricultural contaminated soil. This strain CG5 was able to grow on 10mg of oxadiazon per l, yielding 5.18+/-0.2 mg of protein biomass mol(-1). GC-MS analyses of the metabolites from oxadiazon catabolism revealed its dehalogenation and degradation to form non-toxic end-products, cells were then immobilized by adsorption on a ceramic support to be used as biocatalysts in herbicide removal biofilm-reactor processes. Seventy-two per cent of the oxadiazon was removed, and the maximum specific substrate uptake rate was 10.63+/-0.5 microg h(-1) mg(-1) prot. A new mathematical model was developed to interpret and predict the behaviour of the bacteria and pollutants in a biofilm-reactor system, to consider biofilm structural and morphological properties.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33382
Oxadiazon, PESTANAL®, analytical standard
C15H18Cl2N2O3