EMAIL THIS PAGE TO A FRIEND
Microbiological research
Isolation and characterization of a cold-induced nonculturable suppression mutant of Vibrio vulnificus.
PMID 16520028
Abstract
The viable but nonculturable (VBNC) suppression mutant formed platable cells at low temperature stress after inoculation in artificial seawater (ASW). Suppression subtractive hybridization was used to identify differentially expressed genes among cDNAs of the VBNC suppression mutant and the wild-type Vibrio vulnificus strain. Glutathione S-transferase was identified as a responsive gene of the VBNC suppression mutant in our assay, and was highly expressed from the VBNC suppression mutant at low temperature stress. Culturability tests revealed that the wild-type cells were sensitive to oxidative stress in the hydrogen peroxide (H(2)O(2)) and to 1-chloro-2,4-dinitrobenzene (CDNB) compared with the VBNC suppression mutant cells. Adding glutathione showed that many wild-type V. vulnificus cells maintained culturability in cold ASW. These results suggest that non-nutritional growth inhibitors, such as peroxide that accumulates at low temperatures, influence VBNC in V. vulnificus cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
342122
1-Methyl-3-nitroguanidine, contains ~25% water
C2H6N4O2